missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Pear-Shaped Centrifuge Tubes

Complies with ASTM D96 and D-1966

Brand:  Pyrex 3440/02M

191.32 GBP valid until 2025-06-13
Use promo code "24123" to get your promotional price.


Product Code. 12632132

  • £257.00 / Pack of 2
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Complies with ASTM D96 and D-1966

  • For the determination of water and sediment in petroleum products
  • Graduated in white enamel: - Lower stem graduated 0mL to 1.5mL in 0.1mL divisions - Upper body graduated 1.5mL to 5mL in 0.5mL divisions and from 5mL to 10mL in 1mL divisions
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ASTM D96, D-1966
Borosilicate Glass
Pear
0 to 1.5 mL in 0.1 mL increments lower stem, 1.5 to 5 mL in 0.5 mL increments and from 5 to 10 mL in 1 mL increments upper body
100 mL
For the determination of water and sediment in petroleum products
157 mm
Centrifuge Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyrex™ Pear-Shaped Centrifuge Tubes > Capacity: 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.