Learn More
Novus Biologicals™ Red Blood Cell (RBC) Lysis Buffer
Applications: Functional
Brand: Novus Biologicals™ NBP2-29442
137.60 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
The Red Blood Cell (RBC) Lysis Buffer is for optimal lysis of erythrocytes in single-cell suspensions of mouse hematopoietic tissues such as spleen and human peripheral blood. This buffer contains ammonium chloride, which lyses red cells with minimal effect on lymphocytes. Nucleated red blood cells are not effectively lysed with ammonium chloride. RBC lysis is not necessary when working with mouse thymus and lymph node. Buffer: Dilute 10X RBC Lysis Buffer to 1X in deionized water before use (1mL of 10X RBC lysis buffer with 9mL of deionized water). Bring to room temperature prior to use. Reconstitute: Solubilize the peptides prior to use by making 5mM PBS stock solutions
Specifications
Functional | |
50 mL | |
4°C Do not freeze. | |
Use in functional reported in scientific literature (PMID: 24371146) |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Product Content Correction
Product Title
Novus Biologicals™ Red Blood Cell (RBC) Lysis Buffer >
Spot an opportunity for improvement?Share a Content Correction