Learn More
Thermo Scientific™ Replacement Seals for Thermo Scientific™ MicroClick and 8 x 8 PCR Strip Rotors
A range of options for secure closure on microcentrifuge rotors to help contain samples.
Brand:  Thermo Scientific™ 75005726
106.30 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
 View more versions of this product
Product Code. 15125263
  • £116.37 / Each
Explore more special offers
Description
Description
A range of options for secure closure on microcentrifuge rotors to help contain samples.
  • O-rings for inner and outer seals
  • ClickSeal lids for optimal biocontainment
  • Available for a variety of sizes and models
TRUSTED_SUSTAINABILITY
Specifications
Specifications
O-ring
MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Replacement Seals for Thermo Scientific™ MicroClick and 8 x 8 PCR Strip Rotors > Replacement O-ring for MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors