Learn More
Invitrogen™ SiteClick™ Antibody Labeling Kits
Generate brightly labeled antibodies for flow cytometry, fluorescence imaging, or western blot detection using SiteClick™ Qdot™ R-PE, and Biotin Antibody Labeling Kits.
Brand: Invitrogen™ S20033
419.52 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Generate perfectly labeled antibodies with the SiteClick R-PE, Qdot, or Biotin Antibody Labeling Kits. Site selectivity is achieved by targeting the carbohydrate domains present on essentially all IgG antibodies, regardless of isotype and host species. Depending upon the label, the resulting SiteClick R-PE, Qdot, or Biotin conjugated antibody can be used in flow cytometry, fluorescence imaging, or Western blot detection. Each kit contains everything needed to label 100 μg of IgG antibody with an easy-to-follow, step-by-step protocol. Achieve highly efficient, site-specific, and reproducible labeling with the SiteClick R-PE, Qdot, or Biotin Antibody Labeling Kits.
Conventional amine or thiol labeling chemistry can result in labeling of the antigen binding domain or compromise the structure of an antibody. SiteClick labeling technology, however, attaches the label under mild conditions to the heavy chains of an IgG antibody, ensuring that the antigen binding domains remain available for binding to your antigen target. Because SiteClick Labeling Kits allow simple and gentle site-selective attachment of detection molecules to heavy chain N-linked glycans — far from the antigen-binding domain — excellent reproducibility is achieved during sequential labelings, as well from antibody to antibody. No harsh reduction steps are required, and antibody labeling is consistent and reproducible each time it is performed.
The SiteClick R-PE, Qdot, or Biotin Antibody labeling workflow consists of three steps:
1. antibody carbohydrate domain modification,
2. azide attachment to the antibody and
3. conjugation with the DIBO-modified label.
This workflow employs copper-free click chemistry to covalently link the label containing the DIBO moiety with the azide-modified antibody. Labeling antibodies with the SiteClick antibody labeling system gives you confidence that the antibody will be labeled the same way every time, with no need for reaction optimization. This means the same number of dyes are attached to each molecule, the antigen binding site is preserved, and the same results are achieved every time.
Important features of the SiteClick Qdot Antibody Labeling Kits:
• Include everything required to label 100 μg of IgG antibody (100–250 μg of IgG antibody with the SiteClick Biotin Antibody Labeling Kit)
• Easy to follow step-by-step protocol that require only a few hours hands-on time to complete.
• Highly efficient, site-specific, reproducible labeling chemistry results in high quality antibody conjugates.
Order Info
Store in refrigerator (2–8°C), desiccate, protect from light, and do not freeze.

Specifications
Contains everything needed to azido modify and label 100 μg of antibody | |
1 kit | |
SiteClick Labeling System | |
Wet Ice | |
Biotin |
Labeling Kit | |
Store the entire kit at 2 to 6°C upon receipt | |
100 to 250 μg | |
SiteClick™ | |
Antibodies |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.