missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sterile Syringes for Single Use

Manual syringe without needle for research purposes only. Suitable for use with syringe filters

Brand:  Fisherbrand™ 14955454

 View more versions of this product

Product Code. 15829152

  • £322.00 / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Description

Description

  • GAMMA sterilization
  • Sterile
  • Individually wrapped in a paper/plastic blister pack

Services

5 Years

Specifications

Specifications

19.20 mm
20 mL
Sterile
Slip Tip
Leur Slip
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Sterile Syringes for Single Use > 20mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.