missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Streptococcal Grouping Kit using Latex Agglutination

Rapidly identify streptococcal groups A, B, C, D, F, and G by latex agglutination using this test kit.

Brand:  Thermo Scientific™ DR0593G

85.31 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 13275599

  • £174.00 / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Rapidly identify streptococcal groups A, B, C, D, F, and G by latex agglutination using Thermo Scientific™ Oxoid™ Streptococcal Grouping Kit. This rapid latex agglutination test is designed to deliver simple, rapid and sensitive results for the identification of streptococcal groups A, B, C, D, F, and G . The test utilizes latex particles coated with group-specific antibodies which will agglutinate in the presence of homologous antigens.

Streptococci are facultatively anaerobic, catalase-negative, Gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis, and necrotizing fasciitis.

Use this rapid, simple and sensitive test for the identification of streptococcal groups A, B, C, D, F, and G.

  • Rapid—results in two minutes
  • Simple—easy to implement
  • Visual—easy-to-read results
  • Sensitive—use of latex technology makes this test more sensitive
  • All reagents available separately

Lancefield showed that the majority of pathogenic streptococci possess specific carbohydrate antigens, which permit the classification of streptococci into groups. These streptococcal group antigens can be extracted from the cells and their presence demonstrated with latex particles previously coated with group-specific antibodies. These latex particles will agglutinate in the presence of homologous antigen, but will remain in smooth suspension in the absence of such antigen. The Oxoid Streptococcal Grouping Kit is such a latex agglutination test for the identification of the streptococcal groups and reagents are provided for groups A, B, C, D, F and G. The use of a enzymatic extraction procedure considerably shortens the time required for antigen extraction and much improves the antigen yield, partially for Group D streptococci.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Streptococcal Grouping Enzyme Extraction Reagent
Enzyme Immunoassay Detection
2 Vials
Reagents for Streptococcus Testing
Group A, B, C, D, F and G
2 x 50 Tests
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Instructions

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6185836 Certificate TypeCertificate of Analysis Date20/02/2025 Catalog NumberDR0593G
Lot Number6158071 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog NumberDR0593G
Lot Number6136502 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog NumberDR0593G
Lot Number6120480 Certificate TypeCertificate of Analysis Date01/11/2024 Catalog NumberDR0593G
Lot Number6120479 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog NumberDR0593G
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Streptococcal Grouping Kit using Latex Agglutination > Extraction Enzyme; 2 vials

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.