Specifications
Pocket Refractometer | |
1.333 to 1.356 | |
Zero calibration with distilled water at 20°C | |
160 g |
0 to 15 Brix | |
DIGIT-015 | |
Measuring low sugar concentrations and low refractive values | |
55 x 80 x 205 mm |