Learn More
Thermo Scientific™ UHPLC Filters
Protects columns without compromising performance.
Brand:  Thermo Scientific™ 22180
177.24 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 10127594
  • £186.93 / Pack of 5
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Sacrificing performance is not an option with Thermo Scientific™ UHPLC filters that protect columns for Hypersil GOLD™ 1.9μm and Syncronis 1.7μm columns. These UHPLC filters provide a low-volume filter cartridge design, maintain peak shape, and ensure minimal efficiency loss through dispersion.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.2 μm
2.1 mm I.D. Replacement Filter Cartridge, 0.2 μm
2.1 mm
Thermo Scientific
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ UHPLC Filters >