missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Ultrafiltration Membrane Discs

For sample concentration in stirred cells

£148.00 - £399.00

Specifications

Filter Type Membrane Filter
Format Ultrafiltration Disc
Sterility Non-sterile
For Use With (Application) For sample concentration in stirred cells
Quantity 10/Pk.
View More Specs

Products 30
Product Code Brand Diameter (Metric) Material (Membrane) Thickness (Metric) MWCO Price Quantity & Availability  
Product Code Brand Diameter (Metric) Material (Membrane) Thickness (Metric) MWCO Price Quantity & Availability  
10022281
View Documents
Sartorius
14629--47------D
47 mm Polysulfone 120 μm 5 kDa
£184.00
Pack of 10
Estimated Shipment: 20-06-2025
to see stock.
Add to basket
 
11720945
View Documents
Sartorius
14629--25------D
25 mm Polysulfone 120 μm 5 kDa
£148.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
15992510
View Documents
Sartorius
14650--76------D
76 mm Polysulfone 120 μm 50 kDa
£398.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
10173751
View Documents
Sartorius
14429--25------D
25 mm Cellulose 180 μm 5 kDa
£148.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
11750945
View Documents
Sartorius
14629--76------D
76 mm Polysulfone 120 μm 5 kDa
£398.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
10123941
View Documents
Sartorius
14659--47------D
47 mm Polysulfone 120 μm 30 kDa
£182.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
11790945
View Documents
Sartorius
14639--76------D
76 mm Polysulfone 120 μm 10 kDa
£398.00
Pack of 10
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
 
10504201
View Documents
Sartorius
14459--25------D
25 mm Cellulose 180 μm 30 kDa
£148.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
11740945
View Documents
Sartorius
14629--63------D
63 mm Polysulfone 120 μm 5 kDa
£284.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
10133801
View Documents
Sartorius
14429--63------D
63 mm Cellulose 180 μm 5 kDa
£284.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
11760945
View Documents
Sartorius
14639--25------D
25 mm Polysulfone 120 μm 10 kDa
£185.00
Pack of 10
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
 
11730955
View Documents
Sartorius
14668--47------D
47 mm PES 120 μm 100 kDa
£182.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
10347261
View Documents
Sartorius
14609--47------D
47 mm Polysulfone 120 μm 1 kDa
£183.00
Pack of 10
Estimated Shipment: 20-06-2025
to see stock.
Add to basket
 
10397681
View Documents
Sartorius
14659--63------D
63 mm Polysulfone 120 μm 30 kDa
£284.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
10337261
View Documents
Sartorius
14429--47------D
47 mm Cellulose 180 μm 5 kDa
£182.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
15982510
View Documents
Sartorius
14650--25------D
25 mm Polysulfone 120 μm 50 kDa
£148.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
10748202
View Documents
Sartorius
14650--47------D
47 mm Polysulfone 120 μm 50 kDa
£182.00
Pack of 10
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
 
10662602
View Documents
Sartorius
14459--76------D
76 mm Cellulose 180 μm 30 kDa
£398.00
Pack of 10
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
 
10439511
View Documents
Sartorius
14439--76------D
76 mm Cellulose 180 μm 10 kDa
£398.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
10574571
View Documents
Sartorius
14459--47------D
47 mm Cellulose 180 μm 30 kDa
£182.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
15922520
View Documents
Sartorius
14679--76------D
76 mm Polysulfone 120 μm 300 kDa
£398.00
Pack of 10
Estimated Shipment: 04-06-2025
to see stock.
Add to basket
 
10052301
View Documents
Sartorius
14459--63------D
63 mm Cellulose 180 μm 30 kDa
£284.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
15962510
View Documents
Sartorius
14609--63------D
63 mm Polysulfone 120 μm 1 kDa
£284.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
10716092
View Documents
Sartorius
14639--47------D
47 mm Polysulfone 120 μm 10 kDa
£184.00
Pack of 10
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
10032421
View Documents
Sartorius
14429--76------D
76 mm Cellulose 180 μm 5 kDa
£398.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
10662392
View Documents
Sartorius
14679--47------D
47 mm Polysulfone 120 μm 300 kDa
£182.00
Pack of 10
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
 
15972510
View Documents
Sartorius
14609--76------D
76 mm Polysulfone 120 μm 1 kDa
£399.00
Pack of 10
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
10245801
View Documents
Sartorius
14439--25------D
25 mm Cellulose 180 μm 10 kDa
£148.00
Pack of 10
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
 
11780945
View Documents
Sartorius
14639--63------D
63 mm Polysulfone 120 μm 10 kDa
£284.00
Pack of 10
Estimated Shipment: 09-06-2025
to see stock.
Add to basket
 
15902520
View Documents
Sartorius
14659--25------D
25 mm Polysulfone 120 μm 30 kDa
£148.00
Pack of 10
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
Description

Description

Sartorius Stedim Biotech offers a wide range of ultrafiltration membrane discs for sample concentration in stirred cells. Membrane disc diameters range from 13 mm to 293 mm, molecular weight cut-offs from 1 to 300 kDa. Different membrane materials are available depending on the application: PES is the standard material with high flux, no hydrophobic or hydrophilic interactions and low fouling characteristics.Hydrosart™, a patented regenerated cellulose membrane, features extremely low protein binding and extended chemical resistance and is preferred for concentration of samples with very low protein content.

  • Wide range of MWCO
  • Wide range of disc diameters
  • Compatible with all stirred cells
Specifications

Specifications

Membrane Filter
Non-sterile
10/Pk.
Ultrafiltration Disc
For sample concentration in stirred cells
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius Ultrafiltration Membrane Discs

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.