missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Vinyl Mini Pictograms Hazardous Labels

Brand:  Brady™ 814018

12.70 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 11303472

  • £16.60 / Pack of 96
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Specifications

Specifications

ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Gloss
96/Pk.
Pipe markers and safety signs
0.061 mm
-18°C to 60°C
DANGER of Cutaneous Sensitivity - inhalation
Black on White
MINI PIC1805-10X10-GHS07/96
Vinyl
Permanent Acrylic
10 mm
200 mm
Diamond
First Aid Sign
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Vinyl Mini Pictograms Hazardous Labels > Danger of Cutaneous Sensitivity - inhalation

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.