missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaflow™ 200 Crossflow Cassettes
BIOPHARMA

Offers a standard for ease of use, reliability and flexibility unrivalled by other filtration systems. Sartorius Vivaflow 200 Crossflow Cassettes are ready to use, reusable laboratory crossflow cassettes for concentrating up to 5L samples. Available in different MWCOs and membrane types for different application needs.

Brand:  Sartorius VF20P2

479.99 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 10449321

  • £741.00 / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Rapid, Efficient Concentration
  • Optimally suited for cell culture supernatant concentration, virus concentration and water concentration
  • Utilizes a 200cm2 PES or Hydrosart membrane for high flow rate
  • Thin-channel, flip-flow recirculation path provides high crossflow velocities with minimum pump speed
  • Fast concentration using one or two cassettes
    • A single module typically concentrates 1L by a factor of 50 in less than 30 minutes
    • Two modules used in parallel concentrates 5L by a factor of 50 in under 75 minutes
  • Near total recovery of the concentrate is achieved with a single rinse
  • A 500ml sealed diafiltration reservoir can be added for rapid and convenient buffer exchange
Convenient and Easy to Use
  • One unit comes with all necessary accessories (except pump and pump head)
    • Includes tubing, pressure indicator, flow restrictor and high pressure pump tubing
    • Required equipment: peristaltic pump with a size 16 pump head
  • Can be easily cleaned and re-used several times
  • Clear housing for process monitoring
  • Ready-to-use, disposable cassettes
  • Easily set up two cassettes to run simultaneously for faster concentration
    • Use a pre-assembled Y connector with the correct tubing sizes and fittings
    • Size 16 tubing is used for connecting the two Vivaflow 200 units
    • Size 15 tubing to connect to the pump, with a size 15 pump head
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number243500367 Certificate TypeCertificate of Quality Date28/01/2025 Catalog NumberVF20P2
Lot Number234800937 Certificate TypeCertificate of Quality Date07/11/2024 Catalog NumberVF20P2
Lot Number234700587 Certificate TypeCertificate of Quality Date02/08/2024 Catalog NumberVF20P2
Lot Number234000327 Certificate TypeCertificate of Quality Date27/06/2024 Catalog NumberVF20P2
Lot Number231300247 Certificate TypeCertificate of Quality Date25/06/2024 Catalog NumberVF20P2
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius Vivaflow™ 200 Crossflow Cassettes > Vivaflow 200 Crossflow Cassette; 30,000 MWCO; PES; Ea.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.