Learn More
Sartorius Vivaflow™ 200 Crossflow Cassettes

Offers a standard for ease of use, reliability and flexibility unrivalled by other filtration systems. Sartorius Vivaflow 200 Crossflow Cassettes are ready to use, reusable laboratory crossflow cassettes for concentrating up to 5L samples. Available in different MWCOs and membrane types for different application needs.
Brand: Sartorius VF20P7
479.99 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Rapid, Efficient Concentration- Optimally suited for cell culture supernatant concentration, virus concentration and water concentration
- Utilizes a 200cm2 PES or Hydrosart membrane for high flow rate
- Thin-channel, flip-flow recirculation path provides high crossflow velocities with minimum pump speed
- Fast concentration using one or two cassettes
- A single module typically concentrates 1L by a factor of 50 in less than 30 minutes
- Two modules used in parallel concentrates 5L by a factor of 50 in under 75 minutes
- Near total recovery of the concentrate is achieved with a single rinse
- A 500ml sealed diafiltration reservoir can be added for rapid and convenient buffer exchange
- One unit comes with all necessary accessories (except pump and pump head)
- Includes tubing, pressure indicator, flow restrictor and high pressure pump tubing
- Required equipment: peristaltic pump with a size 16 pump head
- Can be easily cleaned and re-used several times
- Clear housing for process monitoring
- Ready-to-use, disposable cassettes
- Easily set up two cassettes to run simultaneously for faster concentration
- Use a pre-assembled Y connector with the correct tubing sizes and fittings
- Size 16 tubing is used for connecting the two Vivaflow 200 units
- Size 15 tubing to connect to the pump, with a size 15 pump head

Specifications
Ultrafiltration | |
Labscale TFF System | |
PES | |
0.2 μm | |
Non-sterile | |
Cell culture supernatant concentration, virus concentration and water concentration | |
60psi | |
1 Each | |
60°C | |
5 L | |
4.96 x 1.49 x 5.43 in. (12.6 x 3.8 x 13.8cm) | |
3.8 cm | |
13.8 cm |
Cassette | |
Acrylic | |
0.2 μm | |
Up to 2.5 L | |
200 cm2 | |
Pressure indicator, flow restrictor and size 16 PVC peristaltic tubing and fittings | |
200 to 400mL/min. | |
140°F | |
Crossflow Cassette | |
5.3 mL | |
12.6 cm | |
1 cm | |
0.04 cm |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
250700697 | Certificate of Quality | 26/03/2025 | VF20P7 |
243600377 | Certificate of Quality | 09/10/2024 | VF20P7 |
233600627 | Certificate of Quality | 25/06/2024 | VF20P7 |
234200917 | Certificate of Quality | 25/06/2024 | VF20P7 |
234700547 | Certificate of Quality | 25/06/2024 | VF20P7 |
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.