Learn More
Gibco™ Water For Injection (WFI) for Cell Culture

High quality water for use as a solvent in the preparation of cell culture media and laboratory reagents.
Brand: Gibco™ A1287301
15.77 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Gibco™ Water for Injection (WFI) for Cell Culture meets the USP monograph for Water for Injection packaged in bulk for commercial use elsewhere
- The water is prepared by distillation and filtration (0.1 μm), with no added substances
- In addition to meeting USP standards, each lot is quality tested for sterility, endotoxin, conductivity, pH, osmolality, and oxidizable substances
- Conductivity, total organic carbon, endotoxin and bioburden are tested daily; metals are tested monthly
- It can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers
- Should be stored at 15° to 30°C

Specifications
4 to 6 | |
Sterile-filtered | |
500 mL | |
Room Temperature |
Cell Culture | |
None | |
Distilled and filtered (0.1 μm) | |
Gibco™ |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2528640 | Certificate of Analysis | 21/02/2025 | ,A1287306,A1287305,A1287302,A1287303,A1287301 |
2415222 | Certificate of Analysis | 21/02/2025 | ,A1287306,A1287305,A1287302,A1287303,A1287301 |
3072714 | Certificate of Analysis | 21/02/2025 | A1287306,A1287305,A1287302,A1287301 |
2441984 | Certificate of Analysis | 21/02/2025 | ,A1287306,A1287305,A1287302,A1287303,A1287301 |
2505451 | Certificate of Analysis | 21/02/2025 | ,A1287306,A1287305,A1287302,A1287303,A1287301 |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
18Hours
8Minutes
26Seconds
36 -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
49Hours
8Minutes
26Seconds
36New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.