Learn More
QIAGEN QIAcard FTA Elute Indicating Micro
For collection, stabilization, processing, transport and archiving of biological samples. 1 sample area per card.
Brand: QIAGEN WB120411
Description
- FTA Elute technology for convenient water based elution of nucleic acids.
- QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
- Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
- Cell lysis on contact, denatures proteins and protects DNA from degradation.
Specifications
Room Temperature | |
Multiplex PCR, Sequencing after PCR amplification, SNP analysis, STR analysis, HLA typing, Whole genome amplification, Quantitative PCR, Biobanking, Pharmacogenomics, Genotyping, Genetic identification, Transgenic detection | |
Indicating FTA Elute Micro Card | |
40 μL |
Collect, process, transport, and store colorless biological samples, such as buccal or saliva samples. With FTA Elute, proteins remain tightly bound while DNA is eluted from the matrix. | |
Inert dye | |
100 Pk. | |
15 to 30 min |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P501-Dispose of contents/container to .