missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Pyrex™ Capillary Tubes

Manufactured from Pyrex™ and sealed at one end. Bel-Art SP Scienceware™ Wilmad™ Pyrex™ Capillary Tubes provide a low cost solution for small volume NMR measurement.

Brand:  Bel-Art™ 66000-0074

 View more versions of this product

Product Code. 15378648

This item is not returnable. View return policy

Description

Description

  • Made of ASTM Type 1 Class A glass
  • Tested at 400 MHz field
Specifications

Specifications

102 mm
Capillary Tube
2.20 mm
8 in.
WG-1364-2.5A-203M
Borosilicate Glass
2.50 mm
400MHz
5 Pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ SP Scienceware™ Pyrex™ Capillary Tubes >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.