Learn More
Thermo Scientific™ Benchtop Coolers

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.
Brand: Thermo Scientific™ DS5114-0012
116.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.
- Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
- Holds 12, 20 or 32 tubes
- Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
- Printed grid on one-way keyed lid for proper orientation and sample identification
- Molded of impact-resistant polycarbonate with locking handle for long-term use
- Stackable to save space

Specifications
125 mm | |
151 mm | |
108 mm | |
5-15/16 x 4-1/4 x 4-15/16 in. (151 x 108 x 125 mm) | |
3 x 4 | |
Gel-filled/clear lid | |
Benchtop Cooler | |
0.2 to 1.5 mL | |
-20°C |
4.94 in. | |
5.31 in. | |
4.25 in. | |
3x4 compartments, -20°C | |
45 min. | |
Polycarbonate | |
Clear | |
12 Cryogenic Vials (0.2 to 1.5 mL) |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1418854 | Certificate of Compliance | 11/01/2025 | DS5114-0012 |
1324634 | Certificate of Compliance | 10/12/2024 | DS5114-0012 |
1305018 | Certificate of Compliance | 10/12/2024 | DS5114-0012 |
1305371 | Certificate of Compliance | 10/12/2024 | DS5114-0012 |
1404522 | Certificate of Compliance | 19/11/2024 | DS5114-0012 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.