Learn More
DWK Life Sciences Kimble Chase™ Borosilicate Glass 3.3 Reusable Test Tubes
Borosilicate Glass 3.3 Reusable Test Tubes are available in various metric diameters and are DIN 12395 certified.
Brand: DWK Life Sciences 73750-16100
Description
Use Kimble™ Borosilicate Glass 3.3 Reusable Test Tubes in the laboratory. They are available in various metric diameters and are DIN 12395 certified.
- Available with or without rims
- Round bottom
- Autoclavable
Specifications
Round Bottom | |
Borosilicate Glass (3.3) | |
16 mm |
100 mm | |
Autoclavable | |
Reusable Test Tube |
Product Suggestions
Customers who viewed this item also viewed