Learn More
BRAND™ PCR 8-Strip Clear Tubes
Manufactured from polypropylene without lubricants or chemical additives that can contaminate reagents and samples. PCR 8-Strip Tubes are RNase-, DNase-, and DNA-free and are compatible with most 24-, 32-, 96-, and 384-well thermal blocks.
Brand: BRAND™ 781323
74.20 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples. Tube strips are available with or without caps and can be ordered separately.- Thin, uniform walls ensure consistent, precise thermal transfer for excellent results.
- Autoclavable
- RNase-, DNase-, and DNA-free.
- Compatible with most 24-, 32-, 96-, and 384-well thermal blocks.
- Available in convenient tube/cap sets.

Specifications
Green | |
Polypropylene | |
Yes | |
No | |
Tube Strip |
0.2 mL | |
Yes | |
Autoclavable | |
Strip |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.