missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Class B SILBERBRAND ETERNA Graduated Pipette

Class B SILBERBRAND ETERNA Graduated Pipette is constructed of AR-GLAS™ with amber graduations.

Brand:  BRAND™ 27007

 View more versions of this product

Product Code. 15693947

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • AR-GLAS™
  • DIN EN ISO 835
  • Calibrated to deliver (TD, Ex)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Yes
B
AR-GLAS
Amber
2 mL
DIN EN ISO 835
To Deliver (TD, Ex.)
Yes
0.02 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Class B SILBERBRAND ETERNA Graduated Pipette > Volume: 2 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.