Learn More
Contec™ PROSAT™ Sterile Low Endotoxin Wipes
Made from 100% knitted polyester Polynit Heatseal and presaturated with a blend of 70% IPA and the highest grade distilled Water for Injection (WFI). Contec™ PROSAT™ Sterile Low Endotoxin Wipes are low in particles and fibers for ideal usage in product contact areas. Each batch of wipes has a guaranteed endotoxin level of less than <1EU per device.
Brand: Contec™ PSWE0002
Additional Details : CAS Number : 7732-18-5 Weight : 17.70000kg
Description
- Cleanroom laundered, 100% knitted interlock polyester wipes
- The sealed edge wipe produces a very low level of particles and fibers and is ideal for use in product contact areas
- Controlled saturation levels greatly reduce solvent usage, VOC’s and waste
- Guaranteed endotoxin levels of <1EU per device
- Small number of wipes per resealable pouch are ideal for sterile use and help minimize waste
- ISO Class 5 cleanroom compatible
Specifications
12 in. | |
12 in. | |
Polyester | |
No-run interlock knit | |
Most critical pharmaceutical cleanrooms | |
70% IPA/ 30% WFI, USP grade |
30 cm | |
30 cm | |
140 | |
12 in. x 12 in. (30.48 cm x 30.48 cm) | |
30 Wipes/Pouch, Double-bagged | |
Sterile |
Safety and Handling
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.