missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Easy Grip Disposable Polystyrene Sterile Bottles
GREENER_CHOICEBIOPHARMA

Use as replacement receivers for Corning Disposable Sterile Filter Systems or as storage bottles. Corning™ Disposable Sterile Bottles are easy to grip and ideal for storage of media, buffers and other aqueous solutions.

Brand:  Corning™ 430518

153.98 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 10124291

  • £213.00 / Pack of 24
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Disposable polystyrene bottles for storage of media, buffers, and other aqueous solutions
  • Corning low profile style has easy grip sides that facilitate handling
  • Plug seal caps (45 mm) provide an airtight seal and help minimize the risk of contamination.
  • Bottles can be used with Corning vacuum filtration systems with 45 mm neck sizes (Cat. Nos. 430626, 430627, 431161, 430049, 430513, 430514,431118, 430015, and 431174).
  • Sterile
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Single-use
Sterile
Orange HDPE plug seal
24/Cs.
24 Case
1 L
Polystyrene
Applications: Storage of media, buffers and other aqueous solutions
4.5 cm
2/Pk.
Graduated Easy Grip Storage Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number35624027 Certificate TypeCertificate of Quality Date31/03/2025 Catalog Number430518
Lot Number32724026 Certificate TypeCertificate of Quality Date26/03/2025 Catalog Number430518
Lot Number29424025 Certificate TypeCertificate of Quality Date19/03/2025 Catalog Number430518
Lot Number28324022 Certificate TypeCertificate of Quality Date15/02/2025 Catalog Number430518
Lot Number27824021 Certificate TypeCertificate of Quality Date15/02/2025 Catalog Number430518
Special Offers

Special Offers

  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Days

    28

    Hours

    18

    Minutes

    13

    Seconds

    44
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
  • Corning Matrigel , Corning Biocoat , Corning CoolProduct , Corning Media Tech , Corning
    Corning Lab Products Offer
    Promo Code24073

    Expires2025-04-25

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Easy Grip Disposable Polystyrene Sterile Bottles > Volume: 1 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.