Learn More
BRAND™ Dispensette™ S Analog-adjustable Bottletop Dispensers

The Dispensette™ S Analog Bottletop Dispensers feature a floating piston design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for general-purpose dispensing applications.
Brand: BRAND™ 4600120
314.99 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Includes: Discharge tube, valve mounting/calibration tool, polypropylene adapters, telescoping filling tube, recirculation tube (optional), performance certificate, operating manual
Description
- New discharge tube, with or without recirculation valve
- New valve system - no additional sealing rings required, facilitates cleaning and preparations for autoclaving
- Simple priming by optimized flow channels
- Less dispensing force - especially with large volume instruments
- Safe volume setting with internal scalloped track (analog-adjustable instruments)
- Pivoted screw cap - does not disturb dispensing
- Easy to calibrate and adjust in order to comply with GLP guidelines
- Easy to dismantle for cleaning
- Replacable filling and discharge valve with safety bulb (locks when discharge tube is not assembled)
- 360° rotatable valve block allows bottle label to face user
- Telescoping filling tube adjusts easily to different size bottles
- DE-M marking
Services
One year

Specifications
0.5% (10ÎĽL) | |
Autoclavable (121°C) | |
Red | |
0.2 to 2 mL | |
Dispensing aggressive reagents, including concentrated bases and acids such as H3PO4, H2SO4 (except e.g., HCl, HNO3 und HF), saline solutions, as well as many organic solvents |
Bottletop Dispenser | |
ISO 9001 | |
Without recirculation valve | |
Analog | |
0.05 mL |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Brand Dispensette S Accessories
-
Brand Dispensette S Cleaning
-
Brand Dispensette S Teaser
-
Brand Dispensette S Startup
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
12Hours
5Minutes
10Seconds
5 -
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
0Hours
18Minutes
49Seconds
54 -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
29Hours
5Minutes
10Seconds
5
Your input is important to us. Please complete this form to provide feedback related to the content on this product.