Learn More
Gibco™ Ham's F-12 Nutrient Mix, GlutaMAX™ Supplement

Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free single-cell plating of Chinese Hamster Ovary (CHO) cells.
Brand: Gibco™ 31765035
38.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free single-cell plating of Chinese Hamster Ovary (CHO) cells. F-12 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including chondrocytes and rat prostate epithelial cells. We offer a variety of Gibco™ F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This Ham's F-12 is modified as follows:
With: GlutaMAX™, Phenol Red
The complete formulation is available.
Compared to other basal media, F-12 contains a wider variety of components, including zinc, putrescine, hypoxanthine, and thymidine. Serum-free growth of CHO cells in F-12 has led to a variety of improved formulations. Gibco™ F-12 with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.
Gibco™ F-12 contains no proteins or growth factors. Therefore, F-12 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-12 uses a sodium bicarbonate buffer system (1.176 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature

Specifications
CHO, COS-7, and rat prostate epithelial cells | |
Animal Origin-free | |
Liquid | |
Sterile-filtered | |
GlutaMAX, Phenol Red, Sodium Pyruvate | |
cGMP-compliant under the ISO 13485 standard | |
500 mL | |
Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
Primary Rat Astrocytes | |
1 X | |
Ham's F-12 Nutrient Mixture | |
Sterile-filtered | |
No HEPES | |
Gibco™, GlutaMAX™ | |
Room Temperature |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2661559 | Certificate of Analysis | 29/01/2025 | 31765035 |
2323435 | Certificate of Analysis | 29/01/2025 | 31765035 |
2276947 | Certificate of Analysis | 29/01/2025 | 31765035 |
2662206 | Certificate of Analysis | 29/01/2025 | 31765035 |
2756275 | Certificate of Analysis | 29/01/2025 | 31765035 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures. Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. For in vitro diagnostic use.