Learn More
DWK Life Sciences Kimble™ Kim-Kap™ Closures
Available in natural (clear) or choice of four colors
Brand:  DWK Life Sciences 73663-16
 View more versions of this product
Product Code. 10451815
  • £145.00 / Pack of 1000
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Available in natural (clear) or choice of four colors
  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Push Cap
Polypropylene
Non-sterile
Reusable
Autoclavable
Green
16 mm Plain-end Culture Tubes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Kim-Kap™ Closures > For O.D.: 16 mm; Color: Green