missing translation for 'onlineSavingsMsg'
Learn More

Kinase Buffer S (5X)

Kinase Buffer S (5X)

Brand:  Invitrogen PV5213

Product Code. 11535386

  • £145.00 / 4mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Kinase buffer that has been specially optimized for use with LRRK2 and LRRK2 mutants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5X
5X Kinase Buffer S may be shipped frozen or at room temperature. It should be stored at room temperature (20-30°C).
Approved for shipment at Room Temperature or on Wet or Dry Ice
4 mL
Liquid
LRRK2 and LRRK2 mutants
Biochemical ADP accumulation
LRRK2 G2019S, LRRK2
Kinase Buffer S
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2878479A Certificate TypeCertificate of Analysis Date01/02/2024 Catalog NumberPV5213
Lot Number2033238A Certificate TypeCertificate of Analysis Date28/06/2023 Catalog NumberPV5213
Lot Number1969190B Certificate TypeCertificate of Analysis Date08/11/2018 Catalog NumberPV5213
Lot Number1969190A Certificate TypeCertificate of Analysis Date28/02/2018 Catalog NumberPV5213
Lot Number1905054B Certificate TypeCertificate of Analysis Date29/01/2018 Catalog NumberPV5213
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kinase Buffer S (5X) > 4mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.