Learn More
Hirschmann™ labopette™ 8-Channel Pipette
Ergonomically shaped manual pipette, effortless operation and tip ejector.
Brand:  Hirschmann™ 9475602
 View more versions of this product
Product Code. 11652269
  • £1070.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Variable volume setting
  • Multi channel
  • With tip ejector and recalibration possibility
  • Autoclavable
Specifications
Specifications
1.6%
Clear, 1.0-250 μL
Autoclavable
Labopette
10 to 100 μL
0.1 μL
1 Each
ISO 8655
Tip ejector
8
Multichannel Pipette with Variable Volume Setting
Calibration device, individual number and weighing protocol
0.6%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hirschmann™ labopette™ 8-Channel Pipette > Volume (Metric) Range: 10-100 μL