Learn More
Eppendorf™ Tube Racks
GREENER_CHOICE
Optimize workflow with Eppendorf™ Tube Racks that are ideal for sample processing, transport, and short- to mid-term sample storage.
Brand:  Eppendorf™ 0030119843
41.79 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 15770159
  • £48.66 / Pack of 2
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stackable within one rack format and also between all rack variants; saves laboratory space when not in use
  • Non-slip silicone feet and a tilt-proof footprint design ensure safe handling
  • Wavelike design: Tube rows are arranged in different heights so that the reaction vessels are easily accessible and clearly identifiable
  • Made of polypropylene (PP) for high chemical and physical resistance
  • Optimized space-saving dimensions due to perfect tube position-to-footprint arrangement without compromising the tube handling
  • High flexibility: holds tubes of different sizes
  • Numbered tube positions support fast and easy orientation and error-free handling
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Test Tube Stand
Polypropylene
Yes
2 Pk.
PCR Tubes
36
Cryogenic Tubes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ Tube Racks > Positions: 36, For Use With: CryoTubes