Learn More
Invitrogen™ Gel Loading Tips, standard, round
For easy and accurate loading of samples
Brand: Invitrogen™ LC1001
59.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Tips are RNase/DNase-free and non-autoclavable. Standard, round can be used for easy and accurate loading of samples onto 1.0mm and 1.mm gels. Loading volume is 0.5 to 200 μL.
Applications:
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Specifications
Contains 1 pack (200/pack) of Standard, Round gel loading tips. | |
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis | |
Gel Loading Tip | |
200/ pkg. | |
Non-sterile |
Standard round Tips | |
Universal Fit | |
No | |
Room Temperature | |
200 μL |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.