missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Sample Reducing Agent (10X)

Ready-to-use reagent to reduce protein samples for protein gel electrophoresis

Brand:  Invitrogen™ NP0004

9.11 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 10424012

  • £15.40 / 250µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price


Includes: NuPAGE Sample Reducing Agent (500mM dithiothreitol)
This item is not returnable. View return policy

Description

Description

NuPAGE Sample Reducing Agent contains 500mM dithiothreitol (DTT) at a ready-to-use, 10X concentration in a stabilized liquid form. Use NuPAGE Sample Reducing Agent when running proteins on NuPAGE Novex Bis-Tris gels to maintain protein samples in a reduced state during electrophoresis.

1D Gel Electrophoresis, 2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NuPAGE™ Sample Reducing Agent (500 mM dithiothreitol) should be stored at 4°C. Guaranteed stable for 6 months unless otherwise specified in the product literature.
Sample Reducing Agent
250μL
500 mM
NuPAGE™
Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3085032 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberNP0004,NP0004_LITL18 PROMO
Lot Number3016241 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberNP0004,NP0004_LITL18 PROMO
Lot Number2466930 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberNP0004,NP0004_LITL18 PROMO
Lot Number2469497 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberNP0004,NP0004_LITL18 PROMO
Lot Number2526351 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberNP0004,NP0004_LITL18 PROMO
View accessory products
Special Offers

Special Offers

  • Multiple Manufacturers
    Fall in love with big savings
    Promo Code: 24107

    Expires2025-03-28

    Days

    18

    Hours

    6

    Minutes

    3

    Seconds

    7
    New Offer
    Shop Now
  • Multiple Manufacturers
    Save on thousands of eligible products.
    Promo Code: 24111

    Expires2025-03-14

    Days

    4

    Hours

    6

    Minutes

    3

    Seconds

    7
    New Offer
    Save Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ NuPAGE™ Sample Reducing Agent (10X) > 250μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.