missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Opti-MEM™ I Reduced Serum Medium
GREENER_CHOICE

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology.

Brand:  Gibco™ 31985070

29.63 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.


36.64 GBP valid until 2025-12-31
Use promo code "24273" to get your promotional price.


 View more versions of this product

Product Code. 15392402

  • £44.45 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology. Opti-MEM™ I medium is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ I medium can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Opti-MEM™ I modifications for a range of cell culture applications.

This Opti-MEM™ I is modified as follows:

With: L-glutamine, Phenol Red

The complete formulation is confidential. For more information, please contact Technical Services.

Using Opti-MEM™ I Medium

Opti-MEM™ I Reduced-Serum Medium is a unique medium that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%. Opti-MEM™ I medium uses a sodium bicarbonate buffer system (2.4 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sp2, AE-1, CHO, BHK-21, and HEK
Human Origin
Liquid
Sterile-filtered
Glutamine, Phenol Red
Gibco™, Opti-MEM™
Room Temperature
Primary Fibroblasts
1 X
Opti-MEM I
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
500 mL
Store at 2–8°C and protect from light.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2962162 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number31985062,31985070
Lot Number3048024 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number31985062,31985070
Lot Number2537156 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number31985062,31985070
Lot Number2522727 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number31985062,31985070
Lot Number2579438 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number31985062,31985070
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Opti-MEM™ I Reduced Serum Medium > Quantity: 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.