Learn More
Thermo Scientific™ PNPP Substrates
Thermo Scientific™ Pierce PNPP Substrates are powders, tablets, solutions and complete kits of p-nitrophenyl phosphate, a colorimetric, soluble substrate of alkaline phosphatase for use in ELISA applications.
Brand: Thermo Scientific™ 37621
155.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
PNPP (p-Nitrophenyl Phosphate, Disodium Sal) is a widely used substrate for detecting alkaline phosphatase in ELISA applications. When alkaline phosphatase and PNPP are reacted, a yellow water-soluble reaction product is formed. This reaction product absorbs light at 405 nm. Pierce PNPP is available in four formats: dry crystalline powder, single-use tablets, a stable substrate solution, and a complete kit containing substrate solution and diethanolamine buffer.
Thermo Scientific Pierce PNPP (p-Nitrophenyl Phosphate, Disodium Salt) is a colorimetric, soluble substrate of alkaline phosphatase for use in ELISA applications. When alkaline phosphatase and PNPP are reacted, a yellow water-soluble reaction product is formed. This reaction product absorbs light at 405 nm.
Features of PNPP (p-nitrophenyl phosphate):
• One-component alkaline phosphatase (AP) substrate
• Absorbance maximum of 405 nm
• Develops a yellow hue in solution upon reaction with AP
• Safer to use—contains a non-mercury, non-azide preservative
The substrate is offered in several formats:
PNPP Substrate
• Powder Form
PNPP Substrate Tablets
• 5 mg tablets with an inert filler as the balance
PNPP Substrate Kit
• PNPP tablets and a Diethanolamine Buffer to yield more than one liter of substrate. The Diethanolamine Substrate Buffer (Cat. No. 34064) is also provided individually as a 5X concentrate.
1-Step™ PNPP Substrate Solution
• Circumvents this time-consuming and variable introducing step by providing a single component PNPP substrate. This substrate is stable for 12 months at 2–8°C and can be stopped with conventional methods.

Specifications
1-Step™ PNPP Substrate Solution | |
100 mL | |
1-Step™ | |
Alkaline Phosphatase | |
Ambient |
ELISA Substrate | |
Colorimetric | |
Chemical Substrate | |
Upon receipt store product at 4°C. |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ZI4450161 | Certificate of Analysis | 15/10/2024 | 37621 |
ZH4228523 | Certificate of Analysis | 31/08/2024 | 37621 |
ZH4228524 | Certificate of Analysis | 31/08/2024 | 37621 |
ZF4228522 | Certificate of Analysis | 19/06/2024 | 37621 |
ZF4228521 | Certificate of Analysis | 19/06/2024 | 37621 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.