missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Polypropylene Graduated Microtubes
GREENER_CHOICE

Use these tubes for safe handling of valuable and toxic samples as well as radioactive substances. Eppendorf™ Polypropylene Graduated Microtubes feature a hinged lid with a small hook that clips around the rim of the tube.

Brand:  Eppendorf™ 0030121597

41.39 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 10241931

  • £44.24 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Use these tubes for safe handling of valuable and toxic samples as well as radioactive substances. EppendorfTM Polypropylene Graduated Microtubes feature a hinged lid with a small hook that clips around the rim of the tube.

  • Biohazardous substances can be removed, free of aerosols, through a thin membrane in the center of the lid with a cannula
  • Lid easily opens with one hand
  • These tubes for 0.5mL, 1.5mL and 2mL have a graduation scale and writing surfaces
  • The 0.5mL tube is optimized for use in PCR* experiments
  • Assorted color packs are a mixture of yellow, red, blue, green and colorless
  • Autoclavable micropestle for re-suspending pellets
  • Can be centrifuged at up to 23,500xgr
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Clear
PCR, Centrifugation
Sterile
Yes
25,000 x g
2 mL
Polypropylene
100Pack
Biopur
Hinged Snap Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf™ Polypropylene Graduated Microtubes > Color: Clear; Capacity: 2mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.