Learn More
Pyrex™ Borosilicate Glass Narrow Neck Round Bottom Boiling Flask
Resistant to chemical corrosion and thermal shock
Brand:  Pyrex 1100/28D
42.85 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10754972
  • £89.80 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Complies with ISO 1773
  • Manufactured from Pyrex™ borosilicate glass which offers excellent resistance to chemical corrosion and thermal shock
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 1773
Borosilicate Glass
Round
43
2000 mL
260 mm
166 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Pyrex™ Borosilicate Glass Narrow Neck Round Bottom Boiling Flask > Capacity: 2000mL