Learn More
Applied Biosystems™ TaqMan™ SNP Genotyping Assay, human
5' nuclease assay to discriminate between two alleles of a specific SNP for use in genotyping studies
Brand:  Applied Biosystems™ 4351380
 View more versions of this product
Product Code. 10176954
  • £962.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Includes: Two unlabeled PCR primers: (forward and reverse primers at 900nM final concentration); One Applied Biosystems VIC™ dye: MGB labeled probe detects the Allele 1 sequence (probes at 200nM final concentration); One Applied Biosystems 6FAM™ dye: MGB labeled probe detects the Allele 2 sequence (probes at 200nM final concentration)
Description
Description
  • Large selection of SNP assays advances validation and screening studies for disease association and population stratification applications
  • Easy-to-use, single-tube format
  • Robust assay design provides highly accurate, reproducible and reliable results
  • Simple workflow enables quick results
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Two unlabeled PCR primers: (forward and reverse primers at 900nM final concentration); One Applied Biosystems VIC™ dye: MGB labeled probe detects the Allele 1 sequence (probes at 200nM final concentration); One Applied Biosystems 6FAM™ dye: MGB labeled probe detects the Allele 2 sequence (probes at 200nM final concentration). Store at -15° to -25°C
L (2400 reactions), made to order
2400 reactions
Single-Tube TaqMan SNP Genotyping Assays
Genotyping
Mouse
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ TaqMan™ SNP Genotyping Assay, human > L (2400 reactions), made to order