Learn More
Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tube
Manufactured from soda lime glass and constructed with light walls. Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tubes are ideal for general mixing and laboratory work.
Brand: Fisherbrand™ FB63106
Description
- Features: rimmed light walls
- Withstands: moderate warming
- Complies to: ISO 4142

Specifications
ISO 4142 | |
Soda Lime Glass | |
Yes |
150 mm | |
100 Pack | |
18 mm |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.