Learn More
Thermo Scientific™ Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates

Round or square CapMats, Sealing Mats, MatCaps for sealing 96-well storage plates, available in autoclavable and piercable formats.
Brand: Thermo Scientific™ AB-0674
92.17 GBP valid until 2025-06-30
Use promo code "25075" to get your promotional price.
Description
Round or square CapMats, Sealing Mats, MatCaps for sealing 96-well storage plates, available in autoclavable and piercable formats.
Thermo Scientific 96-well Sealing Mats are designed for 96-well storage plates and are available in either piercable or autoclavable formats. The round or square capped mats are made of thermoplastic elastomer and are suitable for long-term storage.
Highlights
- Certified RNAse, DNAse, PCR Inhibitor, and human DNA free
- Round or square capped mats for sealing 96-well storage plates
- Alphanumeric grid-referencing
- Molded from a proprietary thermoplastic elastomer for a flexible design
- Suitable for long term storage
- Available in autoclavable or piercable formats
Pierceable version:
- Solvent resistant (including DMSO, Ethanol, and Isopropanol)
- Suitable for Gamma Irradiation
- Requires 6N for piercing
- Temperature Range: −20°C to +55°C
Autoclavable version:
- Suitable for Gamma Irradiation
- Temperature Range: −20°C to +130°C
Hazardous: No
Storage Condition: Ambient

Specifications
Natural | |
CapMat for 96-well Polypropylene Sample Processing & Storage Plates | |
TPE | |
Autoclavable | |
50 | |
7mm | |
Designed for sealing 96-well storage plate and suitable for long term storage | |
96-well capmat, sealing mat | |
None | |
No | |
Round | |
PCR Ready |
Alcohol Resistant, Solvent Resistant | |
Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant | |
Cleanroom | |
96 | |
10 | |
-4°F to 266°F | |
96-well | |
Abgene | |
Yes | |
-20°C to +130°C | |
Abgene 96 Round Well Plate |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
8Hours
17Minutes
15Seconds
40 -
Thermo Scientific Matrix
Exclusive Discounts on Storage Solutions: Storage Tubes, Equipment and Accessories
Upgrade your lab with top-quality storage consumables and equipment at unbeatable prices.Promo Code: 25075Expires: 2025-06-30
Days
69Hours
17Minutes
15Seconds
40
Your input is important to us. Please complete this form to provide feedback related to the content on this product.