Learn More
Invitrogen™ TRIzol™ Reagent
TRIzol™ Reagent is a complete, ready-to-use reagent for the isolation of high-quality total RNA or the simultaneous isolation of RNA, DNA, and protein from a variety of biological samples.
Brand: Invitrogen™ 15596026
Description
TRIzol™ Reagent is a complete, ready-to-use reagent for the isolation of high-quality total RNA or the simultaneous isolation of RNA, DNA, and protein from a variety of biological samples. This monophasic solution of phenol and guanidine isothiocyanate is designed to isolate separate fractions of RNA, DNA, and proteins from cell and tissue samples of human, animal, plant, yeast, or bacterial origin, within one hour.
Key features of TRIzol™ Reagent include:
• Permits the isolation of RNA, DNA, and protein from the same sample
• Offers superior lysis capability, even with difficult sample types
• Optimized formulations and protocols for tissues, cells, serum, virus, and bacteria
Reliably purify RNA from multiple sample volumes and sources
TRIzol™ Reagent performs well with small quantities of tissue (50–100 mg) and cells (5 × 106), as well as with large quantities of tissue (≥1 g) and cells (>107), and comes with prototcols for purification from samples of human, animal, plant, or bacterial origin. TRIzol™ Reagent maintains the integrity of the RNA due to highly effective inhibition of RNase activity while disrupting cells and dissolving cell components during sample homogenization. The simplicity of the TRIzol™ Reagent method allows simultaneous processing of a large number of samples. The entire procedure can be completed in 1 hour. Total RNA isolated by TRIzol™ Reagent is free of protein and DNA contamination.
Formulated for isolation of multiple molecular targets
TRIzol™ Reagent allows you to perform sequential precipitation of RNA, DNA, and proteins from a single sample. After homogenizing the sample with TRIzol™ Reagent, chloroform is added, and the homogenate is allowed to separate into a clear upper aqueous layer (containing RNA), and interphase and red lower organic layers (containing the DNA and proteins). RNA is precipitated from the aqueous layer with isopropanol. DNA is precipitated from the interphase/organic layer with ethanol. Protein is precipitated from the phenol-ethanol supernatant by isopropanol precipitation. The precipitated RNA, DNA, or protein is washed to remove impurities, and then resuspended for use in downstream applications.
Order Info
Shipping Condition: Room temperature

Specifications
100 mL TRIzol Reagent; room temperature |
|
1 hr. | |
Bacteria, Cells, Plants, Tissue, Yeast | |
Not High-throughput Compatible (Manual) | |
100 mL | |
Bacteria: ≤107 cells Cells: ≤107 Plant: ≤107 cells, 100 mg Tissue: ≤100 mg Yeast: ≤107 cells |
Organic Extraction | |
20 to 600 μL | |
Total RNA, DNA, Protein | |
RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern/Southern/Western blotting, in vitro translation, nuclease protection assays, nucleic acid labeling, hybridization, enzymatic assays, immunoprecipitation, gel shift assays, 2D gel electrophoresis | |
Room Temperature | |
DNA: ≤7 μg RNA: ≤73 μg Protein: varies |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- TRIZOL REAGENT
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- Germ cell mutagenicity Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Phenol, Thiocyanic acid, compound with guanidine (1:1), Ammonium thiocyanate
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
542509 | Certificate of Analysis | 31/01/2025 | 15596026,15596026_LITL18 PROMO |
542506 | Certificate of Analysis | 31/01/2025 | 15596026,15596026_LITL18 PROMO |
542502 | Certificate of Analysis | 31/01/2025 | 15596026,15596026_LITL18 PROMO |
542507 | Certificate of Analysis | 31/01/2025 | 15596026,15596026_LITL18 PROMO |
542505 | Certificate of Analysis | 31/01/2025 | 15596026,15596026_LITL18 PROMO |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.