Learn More
Proteintech CoraLite™488-Phalloidin (green)
Phalloidin is a toxin isolated from mushrooms that binds F-Actin in the cytoskeleton.
Brand:  Proteintech PF00001-300T
Product Code. 17511176
  • £125.00 / 300 tests
Explore more special offers
Description
Description
  • This product is conjugated to our fluorescent CoraLite(R) dye to stain F-actin for labeling, identifying and quantifying actin filaments in immunostaining studies.
  • Compared to many fluorescent dyes, CoraLite(R) dyes have advantages in brightness, light stability and water solubility.
  • Excitation/Emission maxima wavelengths are 490 nm/515 nm
Specifications
Specifications
Immunofluorescence
300 Tests
515 nm
Product Suggestions
Product Suggestions
Videos
SDS Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Proteintech CoraLite™488-Phalloidin (green) > 300 Tests