missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Flat Bottom Cell Culture Plates
GREENER_CHOICEBIOPHARMA

Tissue culture-treated plates for superior cell attachment. Corning™ Costar™ Flat Bottom Cell Culture Microplates provide an excellent device for diagnostic testing when cell adherence is essential.

Brand:  Corning™ 3548

310.87 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


Product Code. 10065370

  • £445.00 / Pack of 100
In Stock
Edge
Explore more special offers
Add to basket

Description

Description

  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications

Specifications

6 x 8
Yes
Flat
Clear
Polystyrene
Sterile
Multi-well Cell Culture Plate
48
100/Cs.
Individually Wrapped
1.13 cm
Clear
Polystyrene
0.28 mL (Well)
0.95 cm2
Rectangular
Cell Culture
0.25 mL
Lid
1/Pk.
100/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number34324007 Certificate TypeCertificate of Quality Date16/04/2025 Catalog Number3548
Lot Number34224017 Certificate TypeCertificate of Quality Date16/04/2025 Catalog Number3548
Lot Number29224039 Certificate TypeCertificate of Quality Date16/04/2025 Catalog Number3548
Lot Number29124040 Certificate TypeCertificate of Quality Date15/04/2025 Catalog Number3548
Lot Number29024025 Certificate TypeCertificate of Quality Date05/04/2025 Catalog Number3548
Special Offers

Special Offers

  • Corning Matrigel , Corning Biocoat , Corning CoolProduct , Corning Media Tech , Corning
    Corning Lab Products Offer
    Promo Code24073

    Expires2025-04-25

  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Days

    13

    Hours

    1

    Minutes

    46

    Seconds

    3
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Costar™ Flat Bottom Cell Culture Plates > 48-well; TC-treated; 1/Pk; 100/Cs

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.