missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Grade 604 Surface Protection Paper
Designed to protect laboratory surfaces against hazardous spills. Fisherbrand™ Grade 604 Surface Protection Paper guards against damage from acids, toxic, corrosive and radioactive fluids and impacts.
Brand: Fisherbrand™ 0604F00021
This item is not returnable.
View return policy
Description
- Available in sheet or roll form
- Polyethylene backed 125g/m2 absorbent paper quickly soaks up spills and prevents flow through to work surface
Specifications
Surface Protection Paper | |
520 mm | |
520 x 420 mm | |
White | |
100 sheets |
Grade 604 Paper | |
420 mm | |
Polyethylene | |
0.25mm | |
Sheet |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 13
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Grade 604 Surface Protection Paper > 520L x 420mmW
Spot an opportunity for improvement?Share a Content Correction