missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Grid inserts for cryo boxes
- 133 x 133 x 30mm (L x W x H)
- white, cardboard
- Available in packs of 10
Specifications
White | |
Square | |
10 | |
133 mm | |
30 mm |
Cardboard | |
Grid Divider | |
133 x 133 x 30 mm | |
133 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Special Offers
- Multiple Manufacturers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Grid Divider >
Spot an opportunity for improvement?Share a Content Correction