Learn More
Invitrogen™ SeeBlue™ Pre-stained Protein Standard
Allows you to easily monitor the progress of your gel run and is an excellent tool for judging Western transfer efficiency
Brand: Invitrogen™ LC5625
80.00 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
108.30 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Includes: 500μL (50 applications of 10μL each) provided in a plastic vial Loading Buffer consists of Tris-HCl, Formamide, SDS, and Phenol Red
Description
The SeeBlue Pre-Stained Standard can be used with NuPAGE™, tris-glycine and tricine gels.
- Nine polypeptides that resolve into sharp, tight blue bands in the range of 4–200 kDa (depending upon the buffer system)
- An excellent indicator for judging Western transfer efficiency
- A ready-to-use format—no mixing, heating, or reducing required
- Gel Compatibility: Bolt Bis-Tris Plus Gels, NovexTricine Gels, NovexTris-Glycine Gels, NuPAGE Bis-Tris Gels
This standard is pre-reduced and does not contain reducing reagents in the loading buffer.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice

Specifications
500 μL (50 applications of 10 μl each) provided in a plastic vial. Loading Buffer consists of Tris-HCl, Formamide, SDS, and Phenol Red. Store at 4°C. |
|
9 | |
Yes | |
1 color: Blue | |
198, 62, 49, 38, 28, 18, 14, 6, 3 kDa | |
500 μL | |
Western Blotting, SDS-PAGE |
Colorimetric | |
Protein Ladder | |
4 to 200 kDa | |
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels | |
SeeBlue™ | |
Wet Ice |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.