missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PTFE Flat Head Stopper
Contains B Length Cone for Europe. Fisherbrand™ PTFE Flat Head Stopper can be used up to 280°C.
Brand: Fisherbrand™ 010.214
Description
- Operates at high vacuum - transverse trapezoidal sealing rings
- Leakage Rate < 1 x 10-4 torr, litre, sec-1
Specifications
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
Ground Glass Joints | |
1/Ea. | |
14/23 | |
Autoclavable | |
11 mm | |
PTFE |
Push Fit | |
34 mm | |
23 x 23 x 34mm | |
-200 to +280°C | |
White | |
23 mm | |
Tapered Stopper |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ PTFE Flat Head Stopper > Joint Size: 14/23
Spot an opportunity for improvement?Share a Content Correction