Learn More
Thermo Scientific™ Sterilin™ Autoclave Bags
Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
Brand: Thermo Scientific™ 510-17
153.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
- HDPE bags are suitable for sterilization at 121°C and polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135°C
- Convenient “tissue box” cartons dispense bags individually
- Coated wire stands or cardboard holders available for use with autoclave bags
Ordering Alerts:
Not available in all geographic regions
Specifications
Autoclave bags (HDPE) | |
500 | |
660 x 305mm | |
660 mm | |
305 mm |
HDPE | |
121°C | |
26 in. | |
12 in. |
Product Suggestions
Customers who viewed this item also viewed