missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 384-Well Plates
GREENER_CHOICEBIOPHARMA

Carry out colormetric assays with reliable and reproducible results using these clear, flat-bottom, immuno nonsterile 384-well plates, offered with a choice of surface.

Brand:  Thermo Scientific™ 464718

191.99 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 10192781

  • £587.00 / Pack of 30
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Alert:

Each lot is functionally tested for IgG binding.


Description

Description

Carry out colormetric assays with reliable and reproducible results using Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 384-Well Plates, offered with a choice of surface for optimal binding. Includes Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.

384-Well Solid Plates:

  • Clear polystyrene
  • Choice of surfaces for optimal binding
  • Alphanumerically labeled rows and columns

Surfaces:

  • Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo ScientificMaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.

Ordering Information:

Please note that availability may vary by country.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

No Barcode
Standard
Colorimetric
30 Cases
Microplate
384
Non-sterile
Square, Flat Bottom (F-well)
10
Antibodies (Glycoproteins)
Hydrophilic
Without Lid
16 x 24
Clear
Polystyrene
100 μL
MaxiSorp
30
None
F384
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1423142 Certificate TypeCertificate of Processing Date18/03/2025 Catalog Number464718
Lot Number1423141 Certificate TypeCertificate of Processing Date11/03/2025 Catalog Number464718
Lot Number1418381 Certificate TypeCertificate of Processing Date21/01/2025 Catalog Number464718
Lot Number1423142 Certificate TypeCertificate of Compliance Date21/02/2025 Catalog Number464718
Lot Number1423141 Certificate TypeCertificate of Compliance Date11/02/2025 Catalog Number464718
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 384-Well Plates > Nunc, Flat-bottom 384, MaxiSorp, Pinchbar, 120μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.