Learn More
Kartell™ 96-Well Microtitration Plates
Manufactured from polystyrene for exceptional strength and durability. Kartell™ 96 Well Microplates, available in a variety of shapes, are an excellent solution for laboratory microtitration needs.
Brand: Kartell™ 02621/S
64.88 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
Manufactured from polystyrene for exceptional strength and durability. Kartell™ 96 Well Microplates, available in a variety of shapes, are an excellent solution for laboratory microtitration needs.
- Aseptic and single wrapped to prevent contamination
- Crystal clear polystyrene assists in preparation visibility
- Polystyrene covers sold separately
- CE marked
- IVD approved
Specifications
Standard | |
Clear | |
96 | |
Round Bottom (U-well) |
8 x 12 | |
Polystyrene | |
Sterile | |
50 |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
8Hours
9Minutes
58Seconds
24