missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ Autoclave Bags
GREENER_CHOICE

Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.

Brand:  Thermo Scientific™ 509L

90.40 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11309103

  • £300.00 / Pack of 200
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.

  • HDPE bags are suitable for sterilization at 121°C and polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135°C
  • Convenient “tissue box” cartons dispense bags individually
  • Coated wire stands or cardboard holders available for use with autoclave bags

Ordering Alerts:

Not available in all geographic regions

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclave bags (HDPE)
200
610 x 406mm
610 mm
406 mm
HDPE
121°C
24 in.
16 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number203974 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number509L
Lot Number80922 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number509L
Lot Number80923 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number509L
Lot Number80621 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number509L
Lot Number86561 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number509L
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Sterilin™ Autoclave Bags >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.