Learn More
Eppendorf™ Tubes™ 3810X

The easy-to-open, securely closing, Eppendorf Tubes™ 3810X offers comfort and dependability for sample preparation, centrifugation, and storage. One vessel for all applications in molecular biology, chemistry, and biochemistry. Safer than ever: designed for centrifugation up to 30,000 x g.
Brand: Eppendorf™ 0030125215
41.99 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Use these tubes for all applications in molecular biology, clinical chemistry and biochemistry. Eppendorf™ 3810X Polypropylene Microcentrifuge Tube offers comfort and dependability for sample preparation, centrifugation and storage.
- Tube lid is easy to open and close for ergonomic operation.
- Polypropylene provides high resistance to chemicals, mechanical stress, and temperature extremes.
- g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples.
- Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
- Graduation for visual control of the pipetted volume.
- Frosted lid and surface on the side for easy labeling.
- Ensured functionality from -86°C to 100°C.
- Autoclavable when open (121°C, 20 min).
- Available in Eppendorf Quality and lot-certified PCR clean purity grade.

Specifications
Autoclavable | |
Clear | |
1000 Pk. | |
-86°C to +100°C | |
PCR Clean | |
0.05 oz. | |
Cylindrical | |
Conical | |
Microcentrifuge Tube |
1.5 mL | |
Polypropylene | |
-122.8°F to +212°F | |
Non-sterile | |
Yes | |
Flat Frost Lid | |
PCR Ready | |
30,000 x g |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
5Hours
5Minutes
23Seconds
14
Your input is important to us. Please complete this form to provide feedback related to the content on this product.